Assay Details
| Gene Symbol: Srpk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009274.2 | NP_033300.2 | 2 - 3 | 401 | 98 | |
| GenBank mRNA | AB006036.1 | - | 2 - 3 | 194 | 98 | |
| AK014004.1 | - | 3 - 4 | 320 | 98 | ||
| AK136861.1 | - | 2 - 3 | 366 | 98 | ||
| AK161384.1 | - | 3 - 4 | 460 | 98 | ||
| AK169580.1 | - | 2 - 3 | 374 | 98 | ||
| BC020178.1 | - | 2 - 3 | 298 | 4507346 | 98 | |
| BC062941.1 | - | 3 - 4 | 346 | 98 | ||
| U92456.1 | - | 3 - 4 | 323 | 98 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC062941.1 transcripts.
Set Membership: |
|

Srpk2
Genomic Map