This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00486452_g1
See other STRA6 GE Assays ›
Gene Symbol:
Stra6
Gene Name:
stimulated by retinoic acid gene 6
Gene Aliases:
AI891933
Chromosome Location:
Chr.9: 58063793 - 58154187 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00486452_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stra6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001162475.1 NP_001155947.1 3 - 4 263 64
NM_001162476.1 NP_001155948.1 3 - 4 574 64
NM_001162479.1 NP_001155951.1 3 - 4 368 64
NM_009291.2 NP_033317.2 3 - 4 533 64
GenBank mRNA AF062476.1 - 3 - 4 533 64
BC075657.1 - 3 - 4 263 30605532 64
CF536552.1 - 3 - 4 351 64
CF730004.1 - 3 - 4 508 64
CF742607.1 - 3 - 4 368 64
CX237908.1 - 1 - 2 60 64

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stra6

Gene Ontology Categories:

Function(s) Process(es)

blood vessel development
kidney development
pulmonary valve morphogenesis
ventricular septum development
transport
heart development
learning
feeding behavior
lung development
adrenal gland development
female genitalia development
retinol transport
vocal learning
ear development
nose morphogenesis
positive regulation of JAK-STAT cascade
lung alveolus development
positive regulation of behavior
digestive tract morphogenesis
embryonic digestive tract development
developmental growth
smooth muscle tissue development
artery morphogenesis
cognition
neuromuscular process
head development
head morphogenesis
face morphogenesis
lung vasculature development
diaphragm development
embryonic camera-type eye formation
eyelid development in camera-type eye
uterus morphogenesis
alveolar primary septum development
pulmonary artery morphogenesis
paramesonephric duct development
ductus arteriosus closure
receptor activity
protein binding
vitamin transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0