This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00486708_g1
See other CAPZB GE Assays ›
Gene Symbol:
Capzb
Gene Name:
capping protein (actin filament) muscle Z-line, beta
Gene Aliases:
1700120C01Rik, AI325129, CPB1, CPB2, CPbeat2, CPbeta1, CPbeta2, Cappb1
Chromosome Location:
Chr.4: 139192899 - 139291820 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00486708_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Capzb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037761.2 NP_001032850.1 5 - 6 601 79
NM_001271405.1 NP_001258334.1 5 - 6 580 79
NM_001271406.1 NP_001258335.1 5 - 6 566 79
NM_009798.4 NP_033928.1 5 - 6 601 79
GenBank mRNA AK153367.1 - 5 - 6 378 79
AK153394.1 - 5 - 6 378 79
AK156778.1 - 5 - 6 570 79
AK159390.1 - 5 - 6 575 79
AK160082.1 - 5 - 6 576 79
AK162757.1 - 5 - 6 568 79
AK168738.1 - 5 - 6 529 79
BC002053.1 - 5 - 6 543 3590980 79
FJ692320.1 - 5 - 6 566 79
U10406.1 - 5 - 6 521 79
U10407.1 - 5 - 6 492 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001741.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Capzb

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein cytoskeletal protein non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
cytoskeleton organization
regulation of lamellipodium assembly
regulation of cell morphogenesis
cell projection organization
lamellipodium assembly
actin cytoskeleton organization
negative regulation of microtubule polymerization
neuron projection development
muscle fiber development
barbed-end actin filament capping
negative regulation of filopodium assembly
actin filament capping
regulation of protein kinase C signaling
actin binding
protein binding
beta-tubulin binding
actin filament binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400014238.mm.1 NM_001037761.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400014238.mm.1 NM_001271405.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400014238.mm.1 NM_009798.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400014238.mm.1 TC0400001741.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400014239.mm.1 NM_001037761.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400014239.mm.1 NM_001271405.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400014239.mm.1 NM_009798.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400014239.mm.1 TC0400001741.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0