This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00486710_m1
See other CAPZB GE Assays ›
Gene Symbol:
Capzb
Gene Name:
capping protein (actin filament) muscle Z-line, beta
Gene Aliases:
1700120C01Rik, AI325129, CPB1, CPB2, CPbeat2, CPbeta1, CPbeta2, Cappb1
Chromosome Location:
Chr.4: 139192899 - 139291820 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00486710_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Capzb
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001037761.2 NP_001032850.1 7 - 8 783 65
NM_001271405.1 NP_001258334.1 7 - 8 762 65
NM_001271406.1 NP_001258335.1 7 - 8 748 65
NM_009798.4 NP_033928.1 7 - 8 783 65
GenBank mRNA AK153367.1 - 7 - 8 560 65
AK153394.1 - 7 - 8 560 65
AK156778.1 - 7 - 8 752 65
AK159390.1 - 7 - 8 757 65
AK162757.1 - 7 - 8 750 65
AK168738.1 - 7 - 8 711 65
AK177260.1 - 2 - 3 111 65
AK177711.1 - 2 - 3 110 65
AK186717.1 - 2 - 3 111 65
AK187793.1 - 2 - 3 111 65
AK188337.1 - 2 - 3 111 65
AK190028.1 - 2 - 3 112 65
AK192076.1 - 2 - 3 111 65
AK207588.1 - 2 - 3 111 65
AK212957.1 - 2 - 3 174 65
AK219201.1 - 2 - 3 111 65
BC002053.1 - 7 - 8 725 3590980 65
FJ692320.1 - 7 - 8 748 65
U10406.1 - 7 - 8 703 65
U10407.1 - 7 - 8 674 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001741.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Capzb

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein cytoskeletal protein non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
cytoskeleton organization
regulation of lamellipodium assembly
regulation of cell morphogenesis
cell projection organization
lamellipodium assembly
actin cytoskeleton organization
negative regulation of microtubule polymerization
neuron projection development
muscle fiber development
barbed-end actin filament capping
negative regulation of filopodium assembly
actin filament capping
regulation of protein kinase C signaling
actin binding
protein binding
beta-tubulin binding
actin filament binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400014242.mm.1 NM_001037761.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400014242.mm.1 NM_001271405.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400014242.mm.1 NM_009798.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400014242.mm.1 TC0400001741.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0