This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00486800_m1
See other CCT4 GE Assays ›
Gene Symbol:
Cct4
Gene Name:
chaperonin containing Tcp1, subunit 4 (delta)
Gene Aliases:
2610204B21Rik, A45, C78323, Cctd
Chromosome Location:
Chr.11: 22990593 - 23003336 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00486800_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cct4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009837.1 NP_033967.1 3 - 4 433 91
GenBank mRNA AK010634.1 - 3 - 4 401 91
AK150601.1 - 3 - 4 380 91
AK152173.1 - 3 - 4 380 91
AK152665.1 - 3 - 4 401 91
AK163369.1 - 3 - 4 388 91
AK165229.1 - 3 - 4 401 91
AK167026.1 - 3 - 4 401 91
AK167262.1 - 3 - 4 370 91
AK167657.1 - 3 - 4 378 91
AK167846.1 - 3 - 4 378 91
AK169339.1 - 3 - 4 380 91
AK200879.1 - 2 - 3 145 91
AK206188.1 - 2 - 3 145 91
AK208716.1 - 2 - 3 91
AK215884.1 - 2 - 3 91
BC054773.1 - 3 - 4 393 6807250 91
L25913.1 - 3 - 4 272 91
Z31554.1 - 3 - 4 433 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000234.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cct4

Panther Classification:

Molecular Function -

chaperonin

Gene Ontology Categories:

Function(s) Process(es)

protein folding
binding of sperm to zona pellucida
toxin transport
nucleotide binding
protein binding
ATP binding
poly(A) RNA binding
unfolded protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100001878.mm.1 NM_009837.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001878.mm.1 NM_009837.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001878.mm.1 TC1100000234.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100001881.mm.1 NM_009837.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001881.mm.1 NM_009837.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001881.mm.1 TC1100000234.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100001880.mm.1 NM_009837.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001880.mm.1 NM_009837.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001880.mm.1 TC1100000234.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100001883.mm.1 NM_009837.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001883.mm.1 NM_009837.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001883.mm.1 TC1100000234.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100001885.mm.1 NM_009837.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001885.mm.1 NM_009837.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001885.mm.1 TC1100000234.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100001877.mm.1 NM_009837.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001877.mm.1 NM_009837.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001877.mm.1 TC1100000234.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100001879.mm.1 NM_009837.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001879.mm.1 NM_009837.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100001879.mm.1 TC1100000234.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-t8jrk:80/100.66.77.131:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0