This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00486817_g1
See other CCT6A GE Assays ›
Gene Symbol:
Cct6a
Gene Name:
chaperonin containing Tcp1, subunit 6a (zeta)
Gene Aliases:
CCT-zeta, CCT-zeta-1, Cct6, Cctz-1, TCP-1-zeta
Chromosome Location:
Chr.5: 129787356 - 129794500 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00486817_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cct6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009838.2 NP_033968.1 958 75
GenBank mRNA AK077916.1 - 912 75
AK134428.1 - 911 75
AK145522.1 - 917 75
AK147130.1 - 955 75
AK149757.1 - 1317 75
AK150977.1 - 912 75
AK159354.1 - 912 75
AK159785.1 - 932 75
AK159975.1 - 958 75
AK160067.1 - 912 75
AK164312.1 - 912 75
AK165612.1 - 914 75
AK166749.1 - 911 75
AK166795.1 - 941 75
AK166870.1 - 909 75
AK166887.1 - 910 75
AK166929.1 - 910 75
AK167007.1 - 940 75
AK167076.1 - 909 75
AK167082.1 - 907 75
AK167164.1 - 917 75
AK167180.1 - 944 75
AK167307.1 - 912 75
AK167669.1 - 941 75
AK167994.1 - 907 75
AK168065.1 - 907 75
AK168940.1 - 941 75
AK169033.1 - 891 75
BC018459.1 - 522 3597455 75
BC055043.1 - 920 75
BC069917.1 - 913 6394714 75
BC094352.1 - 884 6809215 75
Z31557.1 - 929 75

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cct6a

Panther Classification:

Molecular Function -

chaperonin

Gene Ontology Categories:

Function(s) Process(es)

protein folding
binding of sperm to zona pellucida
toxin transport
nucleotide binding
protein binding
ATP binding
poly(A) RNA binding
unfolded protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline