This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00486822_g1
See other CCT6A GE Assays ›
Gene Symbol:
Cct6a BC004004
Gene Name:
chaperonin containing Tcp1, subunit 6a (zeta)
cDNA sequence BC004004
Gene Aliases:
CCT-zeta, CCT-zeta-1, Cct6, Cctz-1, TCP-1-zeta
2400006G15Rik
Chromosome Location:
Chr.5: 129787356 - 129794500 on Build GRCm38
Chromosome Location:
Chr.17: 29268788 - 29302887 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
Assay ID Mm00486822_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cct6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009838.2 NP_033968.1 1535 71
GenBank mRNA AK077916.1 - 1489 71
AK134428.1 - 1488 71
AK145522.1 - 1495 71
AK147130.1 - 1532 71
AK149757.1 - 1894 71
AK150977.1 - 1489 71
AK159354.1 - 1490 71
AK159785.1 - 1509 71
AK159975.1 - 1535 71
AK160067.1 - 1490 71
AK164312.1 - 1489 71
AK165612.1 - 1491 71
AK166749.1 - 1489 71
AK166795.1 - 1519 71
AK166870.1 - 1487 71
AK166887.1 - 1488 71
AK166929.1 - 1487 71
AK167007.1 - 1517 71
AK167076.1 - 1486 71
AK167082.1 - 1485 71
AK167164.1 - 1494 71
AK167180.1 - 1521 71
AK167307.1 - 1489 71
AK167851.1 - 1490 71
AK167994.1 - 1484 71
AK168065.1 - 1484 71
AK168940.1 - 1518 71
AK169033.1 - 1468 71
AK177569.1 - 1 - 2 34 71
AK194557.1 - 1 - 2 71
AK202658.1 - 1 - 2 34 71
AK204480.1 - 1 - 2 34 71
AK219945.1 - 1 - 2 34 71
BC018459.1 - 1099 3597455 71
BC055043.1 - 1497 71
BC069917.1 - 1490 6394714 71
BC094352.1 - 1461 6809215 71
Z31557.1 - 1506 71
Gene Symbol: BC004004
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK180865.1 - 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001512.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cct6a
Click here to find siRNAs for Gene BC004004

Panther Classification:

Molecular Function -

chaperonin

Gene Ontology Categories:

Function(s) Process(es)

protein folding
binding of sperm to zona pellucida
toxin transport
nucleotide binding
protein binding
ATP binding
poly(A) RNA binding
unfolded protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500013687.mm.1 NM_009838.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500013687.mm.1 TC0500001512.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500013686.mm.1 NM_009838.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500013686.mm.1 TC0500001512.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline