This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00486829_m1
See other CCT6B GE Assays ›
Gene Symbol:
Cct6b
Gene Name:
chaperonin containing Tcp1, subunit 6b (zeta)
Gene Aliases:
CCT-zeta-2, CCTzeta-2, Cctz-2, TCP-1-zeta-2
Chromosome Location:
Chr.11: 82719248 - 82764321 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00486829_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cct6b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291242.1 NP_001278171.1 7 - 8 1041 95
NM_009839.3 NP_033969.2 7 - 8 972 95
GenBank mRNA AK076639.1 - 7 - 8 1042 95
BC100457.1 - 7 - 8 939 30918851 95
Z50192.1 - 7 - 8 957 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003366.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cct6b

Panther Classification:

Molecular Function -

chaperonin

Gene Ontology Categories:

Function(s) Process(es)

protein folding
toxin transport
nucleotide binding
molecular_function
ATP binding
unfolded protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100030880.mm.1 NM_009839.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100030880.mm.1 TC1100003366.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0