This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00486846_g1
See other CCT8 GE Assays ›
Gene Symbol:
Cct8
Gene Name:
chaperonin containing Tcp1, subunit 8 (theta)
Gene Aliases:
AI132397, Cctq, Tcpq
Chromosome Location:
Chr.16: 87483325 - 87495869 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00486846_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cct8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009840.3 NP_033970.3 12 - 13 1367 105
GenBank mRNA AK010882.1 - 12 - 13 1368 105
AK014005.1 - 5 - 6 433 105
AK075711.1 - 12 - 13 1348 105
AK075880.1 - 12 - 13 1356 105
AK145661.1 - 12 - 13 1351 105
AK145754.1 - 12 - 13 1350 105
AK145918.1 - 12 - 13 1369 105
AK147038.1 - 12 - 13 1370 105
AK159969.1 - 12 - 13 1358 105
AK172867.1 - 12 - 13 1338 105
BC009007.1 - 12 - 13 1325 3582639 105
Z37164.1 - 12 - 13 1371 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001986.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cct8

Panther Classification:

Molecular Function -

chaperonin

Gene Ontology Categories:

Function(s) Process(es)

protein folding
binding of sperm to zona pellucida
pore complex assembly
toxin transport
nucleotide binding
protein binding
ATP binding
unfolded protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600015423.mm.1 NM_009840.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600015423.mm.1 NM_009840.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600015423.mm.1 TC1600001986.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline