This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00486979_g1
See other CEL GE Assays ›
Gene Symbol:
Cel
Gene Name:
carboxyl ester lipase
Gene Aliases:
1810036E18Rik, BAL
Chromosome Location:
Chr.2: 28555819 - 28563362 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00486979_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cel
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009885.1 NP_034015.1 5 - 6 666 117
GenBank mRNA AK131818.1 - 5 - 6 684 117
AK131824.1 - 5 - 6 684 117
BC006872.1 - 5 - 6 678 2646703 117
U33169.1 - 5 - 6 666 117
U37386.1 - 5 - 6 670 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003232.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cel

Panther Classification:

Molecular Function -

cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
cholesterol catabolic process
lipid catabolic process
pancreatic juice secretion
retinol metabolic process
ceramide catabolic process
lysophospholipase activity
sterol esterase activity
triglyceride lipase activity
hydrolase activity
protein complex binding
glycosphingolipid binding
retinyl-palmitate esterase activity
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200027238.mm.1 NM_009885.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200027238.mm.1 TC0200003232.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0