Assay Details
| Gene Symbol: Gamt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010255.3 | NP_034385.1 | 2 - 3 | 392 | 75 | |
| GenBank mRNA | AA881765.1 | - | 2 - 3 | 73 | ||
| AF015887.1 | - | 2 - 3 | 325 | 75 | ||
| AK140688.1 | - | 2 - 3 | 393 | 75 | ||
| AK158087.1 | - | 2 - 3 | 335 | 75 | ||
| BC049233.1 | - | 2 - 3 | 378 | 4195030 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gamt
Genomic Map