This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00487985_g1
See other PITPNM2 GE Assays ›
Gene Symbol:
Pitpnm2
Gene Name:
phosphatidylinositol transfer protein, membrane-associated 2
Gene Aliases:
NIR3, RDGBA2, Rdgb2, mKIAA1457
Chromosome Location:
Chr.5: 124118689 - 124249760 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00487985_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pitpnm2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289472.1 NP_001276401.1 8 - 9 1483 88
NM_011256.3 NP_035386.1 9 - 10 1674 88
GenBank mRNA AF058693.1 - 8 - 9 1342 88
AK015126.1 - 8 - 9 1432 88
AK053524.1 - 3 - 4 4358 88
AK129364.1 - 8 - 9 1483 88
BC070452.1 - 8 - 9 1483 30621943 88
BC082303.1 - 9 - 10 1674 88

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pitpnm2

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
phospholipid binding
lipid binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline