This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00488465_m1
See other STAU1 GE Assays ›
Gene Symbol:
Stau1
Gene Name:
staufen (RNA binding protein) homolog 1 (Drosophila)
Gene Aliases:
5830401L18Rik, AW549911, C85792, Stau
Chromosome Location:
Chr.2: 166947549 - 166996299 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00488465_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stau1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109905.2 NP_001103375.1 7 - 8 1166 109
NM_001109906.2 NP_001103376.1 7 - 8 1184 109
NM_001291160.1 NP_001278089.1 7 - 8 1166 109
NM_011490.4 NP_035620.1 7 - 8 1160 109
NR_111895.1 6 - 7 918 109
GenBank mRNA AF061942.1 - 7 - 8 1160 109
AF395842.1 - 6 - 7 887 109
AK005002.2 - 7 - 8 1107 109
AK139532.1 - 7 - 8 1113 109
AK145826.1 - 7 - 8 1151 109
AK151835.1 - 7 - 8 972 109
AK152269.1 - 7 - 8 978 109
AK152744.1 - 7 - 8 972 109
AK159602.1 - 7 - 8 1101 109
AK159781.1 - 7 - 8 1136 109
AK160979.1 - 7 - 8 1125 109
AK167637.1 - 7 - 8 1107 109
AK194004.1 - 2 - 3 163 109
AK206904.1 - 2 - 3 163 109
AY391773.1 - 6 - 7 917 109
BC012959.1 - 7 - 8 1130 4188394 109
BC082277.1 - 7 - 8 1103 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200005179.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stau1

Gene Ontology Categories:

Function(s) Process(es)

intracellular mRNA localization
positive regulation of viral genome replication
positive regulation by virus of viral protein levels in host cell
RNA binding
double-stranded RNA binding
protein binding
protein phosphatase 1 binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200044302.mm.1 NM_001109905.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200044302.mm.1 NM_001109906.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200044302.mm.1 NM_011490.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200044302.mm.1 TC0200005179.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200044300.mm.1 NM_001109905.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200044300.mm.1 NM_001109906.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200044300.mm.1 NM_011490.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200044300.mm.1 TC0200005179.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0