This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00488468_g1
See other STAU1 GE Assays ›
Gene Symbol:
Stau1
Gene Name:
staufen (RNA binding protein) homolog 1 (Drosophila)
Gene Aliases:
5830401L18Rik, AW549911, C85792, Stau
Chromosome Location:
Chr.2: 166947549 - 166996299 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00488468_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stau1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109905.2 NP_001103375.1 10 - 11 1697 78
NM_001109906.2 NP_001103376.1 10 - 11 1715 78
NM_001291160.1 NP_001278089.1 10 - 11 1697 78
NM_011490.4 NP_035620.1 10 - 11 1691 78
NR_111895.1 10 - 11 1364 78
GenBank mRNA AF061942.1 - 10 - 11 1691 78
AF395842.1 - 9 - 10 1418 78
AK005002.2 - 10 - 11 1638 78
AK139532.1 - 10 - 11 1644 78
AK145826.1 - 10 - 11 1682 78
AK151835.1 - 10 - 11 1503 78
AK152269.1 - 10 - 11 1509 78
AK152744.1 - 10 - 11 1503 78
AK158952.1 - 5 - 6 2278 78
AK159602.1 - 10 - 11 1632 78
AK159781.1 - 10 - 11 1667 78
AK160979.1 - 10 - 11 1656 78
AK167637.1 - 10 - 11 1638 78
AK177429.1 - 1 - 2 57 78
AY391773.1 - 10 - 11 1363 78
BC012959.1 - 10 - 11 1661 4188394 78
BC082277.1 - 10 - 11 1634 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200005179.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stau1

Gene Ontology Categories:

Function(s) Process(es)

intracellular mRNA localization
positive regulation of viral genome replication
positive regulation by virus of viral protein levels in host cell
RNA binding
double-stranded RNA binding
protein binding
protein phosphatase 1 binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200044294.mm.1 NM_001109905.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200044294.mm.1 NM_001109906.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200044294.mm.1 NM_011490.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200044294.mm.1 TC0200005179.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0