This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00488587_m1
See other CLPX GE Assays ›
Gene Symbol:
Clpx
Gene Name:
caseinolytic mitochondrial matrix peptidase chaperone subunit
Gene Aliases:
AU014732, E330029I21
Chromosome Location:
Chr.9: 65294250 - 65330658 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00488587_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Clpx
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001044389.2 NP_001037854.1 4 - 5 738 82
NM_011802.3 NP_035932.2 4 - 5 738 82
GenBank mRNA AF134983.1 - 4 - 5 689 82
AJ276991.1 - 4 - 5 620 82
AK087848.1 - 4 - 5 692 82
BC061153.1 - 4 - 5 696 6593118 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000835.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Clpx

Panther Classification:

Molecular Function -

protease

Gene Ontology Categories:

Function(s) Process(es)

protein folding
positive regulation of peptidase activity
proteolysis involved in cellular protein catabolic process
nucleotide binding
ATP-dependent peptidase activity
protein binding
ATP binding
peptidase activator activity
ATPase activity
metal ion binding
unfolded protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900006377.mm.1 NM_001044389.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900006377.mm.1 NM_011802.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006377.mm.1 TC0900000835.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900006378.mm.1 NM_011802.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900006378.mm.1 TC0900000835.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0