This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00488841_m1
See other RACGAP1 GE Assays ›
Gene Symbol:
Racgap1
Gene Name:
Rac GTPase-activating protein 1
Gene Aliases:
AI227039, AI327394, Band25, GTPase, MgcRacGAP, gtl11, mKIAA1478
Chromosome Location:
Chr.15: 99620496 - 99651656 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00488841_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Racgap1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253808.1 NP_001240737.1 4 - 5 579 112
NM_001253809.1 NP_001240738.1 4 - 5 575 112
NM_012025.7 NP_036155.1 5 - 6 710 112
GenBank mRNA AB030252.1 - 4 - 5 559 112
AF079974.1 - 4 - 5 520 112
AF212320.1 - 4 - 5 546 112
AF212321.1 - 5 - 6 677 112
AK053850.1 - 4 - 5 540 112
AK144608.1 - 5 - 6 678 112
AK154929.1 - 5 - 6 698 112
AK165897.1 - 4 - 5 532 112
AK166084.1 - 5 - 6 668 112
AK166094.1 - 4 - 5 537 112
AK168025.1 - 4 - 5 536 112
AK168170.1 - 4 - 5 536 112
AK168679.1 - 4 - 5 520 112
AK183940.1 - 2 - 3 170 112
AK207314.1 - 2 - 3 170 112
BC010715.1 - 4 - 5 525 3602242 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002217.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Racgap1

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

mitotic cytokinesis
actomyosin contractile ring assembly
transport
ion transport
cell cycle
signal transduction
multicellular organism development
spermatogenesis
sulfate transport
embryo development
cell differentiation
positive regulation of cytokinesis
intracellular signal transduction
positive regulation of GTPase activity
mitotic spindle midzone assembly
cell division
regulation of attachment of spindle microtubules to kinetochore
mitotic spindle assembly
GTPase activator activity
phosphatidylinositol-3,4,5-trisphosphate binding
microtubule binding
lipid binding
protein kinase binding
alpha-tubulin binding
gamma-tubulin binding
metal ion binding
beta-tubulin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500018181.mm.1 NM_001253808.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018181.mm.1 NM_001253808.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018181.mm.1 NM_001253809.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018181.mm.1 NM_001253809.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018181.mm.1 NM_012025.7.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018181.mm.1 NM_012025.7 Refseq
Affymetrix Mouse Clariom S Array PSR1500018181.mm.1 TC1500002217.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500018184.mm.1 NM_001253808.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018184.mm.1 NM_001253808.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018184.mm.1 NM_001253809.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018184.mm.1 NM_001253809.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018184.mm.1 NM_012025.7.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018184.mm.1 NM_012025.7 Refseq
Affymetrix Mouse Clariom S Array PSR1500018184.mm.1 TC1500002217.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0