This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00488970_m1
See other MEF2A GE Assays ›
Gene Symbol:
Mef2a
Gene Name:
myocyte enhancer factor 2A
Gene Aliases:
A430079H05Rik
Chromosome Location:
Chr.7: 67231163 - 67373501 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00488970_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mef2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033713.2 NP_001028885.1 10 - 11 1342 64
NM_001291191.1 NP_001278120.1 9 - 10 1344 64
NM_001291192.1 NP_001278121.1 9 - 10 1344 64
NM_001291195.1 NP_001278124.1 9 - 10 1318 64
NM_001291196.1 NP_001278125.1 10 - 11 1465 64
GenBank mRNA AK040227.1 - 5 - 6 1240 64
AK052385.1 - 10 - 11 1466 64
AK132678.1 - 9 - 10 1200 64
AK153751.1 - 9 - 10 1345 64
BC061128.1 - 10 - 11 1313 6593862 64
BC096598.1 - 9 - 10 1286 4979487 64
U30823.1 - 8 - 9 1091 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003442.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mef2a

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial genome maintenance
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
apoptotic process
multicellular organism development
nervous system development
cell differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
mitochondrion distribution
dendrite morphogenesis
ventricular cardiac myofibril assembly
cardiac conduction
ERK5 cascade
cellular response to calcium ion
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
RNA polymerase II transcription coactivator activity
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
protein kinase binding
activating transcription factor binding
histone acetyltransferase binding
histone deacetylase binding
sequence-specific DNA binding
SMAD binding
protein heterodimerization activity
protein dimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700029374.mm.1 NM_001033713.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700029374.mm.1 TC0700003442.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0