This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00489430_m1
See other SH3BGR GE Assays ›
Gene Symbol:
Sh3bgr
Gene Name:
SH3-binding domain glutamic acid-rich protein
Gene Aliases:
5430437A18Rik
Chromosome Location:
Chr.16: 96200470 - 96228937 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00489430_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sh3bgr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015825.2 NP_056640.1 3 - 4 548 113
GenBank mRNA AJ239082.1 - 3 - 4 314 113
AJ272170.1 - 3 - 4 376 113
BC055870.1 - 3 - 4 365 113

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001080.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sh3bgr

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
SH3 domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600008552.mm.1 NM_015825.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600008552.mm.1 NM_015825.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600008552.mm.1 TC1600001080.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-7wltc:80/100.66.72.81:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline