This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00489568_m1
See other SCLY GE Assays ›
Gene Symbol:
Scly
Gene Name:
selenocysteine lyase
Gene Aliases:
9830169H08, A930015N15Rik, SCL, Scly1, Scly2
Chromosome Location:
Chr.1: 91298295 - 91321080 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00489568_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Scly
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016717.3 NP_057926.2 8 - 9 948 73
GenBank mRNA AF175407.1 - 8 - 9 900 73
AK036750.1 - 8 - 9 950 73
AK206313.1 - 2 - 3 133 73
BC019879.1 - 8 - 9 908 5123843 73
BC021389.1 - 8 - 9 932 5054691 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000892.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Scly

Panther Classification:

Molecular Function -

lyase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

selenium compound metabolic process
lipid metabolic process
metabolic process
response to insulin
negative regulation of cellular response to oxidative stress
catalytic activity
selenocysteine lyase activity
transferase activity
lyase activity
pyridoxal phosphate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100007563.mm.1 NM_016717.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100007563.mm.1 NM_016717.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100007563.mm.1 TC0100000892.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100007564.mm.1 NM_016717.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100007564.mm.1 NM_016717.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100007564.mm.1 TC0100000892.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8mgvv:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0