This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00489652_m1
See other FOLH1 GE Assays ›
Gene Symbol:
Folh1
Gene Name:
folate hydrolase 1
Gene Aliases:
GCP2, mopsm
Chromosome Location:
Chr.7: 86718781 - 86776055 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00489652_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Folh1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159706.1 NP_001153178.1 10 - 11 1318 79
NM_016770.3 NP_058050.3 11 - 12 1417 79
GenBank mRNA AF026380.1 - 11 - 12 1399 79
AK002920.1 - 11 - 12 1384 79
AK090289.1 - 1419 79
BC119604.1 - 10 - 11 1262 40060385 79
BC119605.1 - 11 - 12 1361 40060386 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003699.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Folh1

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
folic acid-containing compound metabolic process
metabolic process
positive regulation of apoptotic process
catalytic activity
carboxypeptidase activity
peptidase activity
metallopeptidase activity
hydrolase activity
dipeptidase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700031251.mm.1 NM_001159706.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700031251.mm.1 NM_001159706.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700031251.mm.1 NM_016770.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700031251.mm.1 NM_016770.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700031251.mm.1 TC0700003699.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline