This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00490601_m1
See other MBTPS1 GE Assays ›
Gene Symbol:
Mbtps1
Gene Name:
membrane-bound transcription factor peptidase, site 1
Gene Aliases:
0610038M03Rik, AV003995, S1P, SKI-1, Ski1, mKIAA0091
Chromosome Location:
Chr.8: 119508151 - 119558825 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00490601_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mbtps1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001167910.1 NP_001161382.1 13 - 14 2227 62
NM_019709.4 NP_062683.3 14 - 15 2376 62
GenBank mRNA AF094820.1 - 12 - 13 1795 62
AK029048.1 - 13 - 14 2204 62
AK122202.1 - 13 - 14 2193 62
AK147336.1 - 12 - 13 4796 62
AK147495.1 - 13 - 14 2201 62
BC011533.1 - 13 - 14 2190 62
BC054837.1 - 12 - 13 1995 6490394 62
BC057198.1 - 14 - 15 2342 5310414 62
DQ913734.1 - 12 - 13 1858 62

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mbtps1

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
lipid metabolic process
lysosome organization
steroid metabolic process
cholesterol metabolic process
regulation of transcription factor import into nucleus
endopeptidase activity
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline