This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00492099_g1
See other DDOST GE Assays ›
Gene Symbol:
Ddost
Gene Name:
dolichyl-di-phosphooligosaccharide-protein glycotransferase
Gene Aliases:
-
Chromosome Location:
Chr.4: 138304738 - 138312611 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00492099_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ddost
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007838.2 NP_031864.2 1 - 2 305 70
GenBank mRNA AK077651.1 - 1 - 2 168 70
AK081542.1 - 1 - 2 197 70
AK084267.1 - 1 - 2 189 70
AK148095.1 - 1 - 2 186 70
AK150683.1 - 1 - 2 196 70
BC068132.1 - 1 - 2 305 6808198 70
D89063.1 - 1 - 2 307 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001722.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ddost

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

protein N-linked glycosylation
protein N-linked glycosylation via asparagine
response to cytokine
T cell activation
dolichyl-diphosphooligosaccharide-protein glycotransferase activity
transferase activity
transferase activity, transferring glycosyl groups

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400014089.mm.1 NM_007838.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400014089.mm.1 NM_007838.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400014089.mm.1 TC0400001722.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400014090.mm.1 NM_007838.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400014090.mm.1 NM_007838.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400014090.mm.1 TC0400001722.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 761885 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0