This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00492103_g1
See other DDOST GE Assays ›
Gene Symbol:
Ddost
Gene Name:
dolichyl-di-phosphooligosaccharide-protein glycotransferase
Gene Aliases:
-
Chromosome Location:
Chr.4: 138304738 - 138312611 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00492103_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ddost
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_007838.2 NP_031864.2 5 - 6 698 76
GenBank mRNA AK077651.1 - 5 - 6 560 76
AK081542.1 - 5 - 6 590 76
AK084267.1 - 5 - 6 582 76
AK148095.1 - 5 - 6 579 76
AK150683.1 - 5 - 6 589 76
AK200087.1 - 3 - 4 259 76
BC068132.1 - 5 - 6 698 6808198 76
D89063.1 - 5 - 6 700 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001722.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ddost

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

protein N-linked glycosylation
protein N-linked glycosylation via asparagine
response to cytokine
T cell activation
dolichyl-diphosphooligosaccharide-protein glycotransferase activity
transferase activity
transferase activity, transferring glycosyl groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400014093.mm.1 NM_007838.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400014093.mm.1 NM_007838.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400014093.mm.1 TC0400001722.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400014094.mm.1 NM_007838.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400014094.mm.1 NM_007838.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400014094.mm.1 TC0400001722.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0