This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00492116_g1
See other DHX15 GE Assays ›
Gene Symbol:
Dhx15
Gene Name:
DEAH (Asp-Glu-Ala-His) box polypeptide 15
Gene Aliases:
DBP1, DEAH9, Ddx15, HRH2, mDEAH9
Chromosome Location:
Chr.5: 52150203 - 52190546 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00492116_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dhx15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042620.2 NP_001036085.1 8 - 9 1682 80
NM_007839.3 NP_031865.2 8 - 9 1682 80
NR_104311.1 8 - 9 1682 80
GenBank mRNA AF017153.1 - 8 - 9 1530 80
AK145983.1 - 8 - 9 1632 80
AK146441.1 - 8 - 9 1629 80
AK149869.1 - 8 - 9 1644 80
AK151015.1 - 8 - 9 1645 80
AK151757.1 - 8 - 9 1645 80
AK167927.1 - 8 - 9 1555 80
AK168835.1 - 8 - 9 1650 80
AK169174.1 - 8 - 9 1650 80
BC003745.1 - 8 - 9 1625 80
BC100343.1 - 8 - 9 1625 30935044 80
BC144759.1 - 8 - 9 1513 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002406.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dhx15

Panther Classification:

Molecular Function -

RNA helicase

Gene Ontology Categories:

Function(s) Process(es)

RNA processing
mRNA processing
RNA splicing
nucleotide binding
nucleic acid binding
double-stranded RNA binding
ATP-dependent RNA helicase activity
helicase activity
protein binding
ATP binding
ATP-dependent helicase activity
hydrolase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500021710.mm.1 NM_001042620.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500021710.mm.1 NM_007839.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500021710.mm.1 NR_104311.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500021710.mm.1 TC0500002406.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline