This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00492640_m1
See other IL1RAP GE Assays ›
Gene Symbol:
Il1rap
Gene Name:
interleukin 1 receptor accessory protein
Gene Aliases:
6430709H04Rik, AI255955, AV239853, IL-1RAcP
Chromosome Location:
Chr.16: 26581688 - 26730128 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00492640_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il1rap
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159317.1 NP_001152789.1 8 - 9 1238 64
NM_001159318.1 NP_001152790.1 8 - 9 1238 64
NM_008364.2 NP_032390.1 8 - 9 1238 64
GenBank mRNA AK033972.1 - 8 - 9 1187 64
AK034269.1 - 8 - 9 1202 64
AK045686.1 - 8 - 9 1187 64
AK083209.1 - 8 - 9 1235 64
AK136782.1 - 8 - 9 1189 64
X85999.1 - 8 - 9 1189 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000376.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Il1rap

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

immune system process
inflammatory response
signal transduction
cytokine-mediated signaling pathway
interleukin-2 biosynthetic process
innate immune response
signal transducer activity
interleukin-1 receptor activity
interleukin-1 receptor binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600003578.mm.1 NM_001159317.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003578.mm.1 NM_001159317.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003578.mm.1 NM_001159318.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003578.mm.1 NM_001159318.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003578.mm.1 NM_008364.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003578.mm.1 NM_008364.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600003578.mm.1 TC1600000376.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600003586.mm.1 NM_001159317.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003586.mm.1 NM_001159317.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003586.mm.1 NM_001159318.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003586.mm.1 NM_001159318.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003586.mm.1 TC1600000376.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600003584.mm.1 NM_001159318.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003584.mm.1 NM_001159318.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003584.mm.1 TC1600000376.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600003585.mm.1 NM_001159317.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003585.mm.1 NM_001159317.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003585.mm.1 NM_001159318.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003585.mm.1 NM_001159318.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003585.mm.1 TC1600000376.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600003577.mm.1 NM_001159317.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003577.mm.1 NM_001159317.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003577.mm.1 NM_001159318.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003577.mm.1 NM_001159318.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003577.mm.1 NM_008364.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003577.mm.1 NM_008364.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600003577.mm.1 TC1600000376.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600003582.mm.1 NM_001159317.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003582.mm.1 NM_001159317.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003582.mm.1 NM_001159318.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003582.mm.1 NM_001159318.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003582.mm.1 TC1600000376.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600003587.mm.1 NM_001159317.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003587.mm.1 NM_001159317.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003587.mm.1 NM_001159318.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003587.mm.1 NM_001159318.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003587.mm.1 TC1600000376.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600003583.mm.1 NM_001159317.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003583.mm.1 NM_001159317.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003583.mm.1 NM_001159318.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003583.mm.1 NM_001159318.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600003583.mm.1 TC1600000376.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mmql6:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0