This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00493448_m1
See other TCF7 GE Assays ›
Gene Symbol:
Tcf7
Gene Name:
transcription factor 7, T cell specific
Gene Aliases:
AI465550, TCF-1, Tcf1
Chromosome Location:
Chr.11: 52252283 - 52283378 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00493448_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tcf7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009331.3 NP_033357.1 5 - 6 442 95
GenBank mRNA AK037616.1 - 5 - 6 443 95
BC056630.1 - 4 - 5 684 95
BC138596.1 - 4 - 5 422 8861616 95
BC145343.1 - 4 - 5 422 9053899 95
X61385.1 - 6 - 7 584 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002715.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tcf7

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
Wnt signaling pathway
neural tube development
embryonic genitalia morphogenesis
T cell receptor V(D)J recombination
regulation of cell proliferation
canonical Wnt signaling pathway involved in negative regulation of apoptotic process
alpha-beta T cell differentiation
embryonic digestive tract morphogenesis
embryonic hindgut morphogenesis
generation of neurons
canonical Wnt signaling pathway
core promoter binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
beta-catenin binding
sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100024740.mm.1 NM_009331.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100024740.mm.1 TC1100002715.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0