This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00494070_m1
See other UCP1 GE Assays ›
Gene Symbol:
Ucp1
Gene Name:
uncoupling protein 1 (mitochondrial, proton carrier)
Gene Aliases:
AI385626, Slc25a7, Ucp
Chromosome Location:
Chr.8: 83290348 - 83298456 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00494070_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ucp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009463.3 NP_033489.1 2 - 3 556 81
GenBank mRNA AK002759.1 - 2 - 3 554 81
AK076454.1 - 2 - 3 553 81
BC012701.1 - 2 - 3 553 4191276 81
U63419.1 - 2 - 3 552 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000919.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ucp1

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
transport
mitochondrial transport
proton transport
transmembrane transport
protein binding
oxidative phosphorylation uncoupler activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800006788.mm.1 NM_009463.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006788.mm.1 NM_009463.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800006788.mm.1 TC0800000919.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0