This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00494247_m1
See other PLAGL1 GE Assays ›
Gene Symbol:
Plagl1
Gene Name:
pleiomorphic adenoma gene-like 1
Gene Aliases:
Lot1, Zac1
Chromosome Location:
Chr.10: 13090788 - 13131694 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00494247_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Plagl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK142052.1 - 1 - 2 196 59
X95503.1 - 1 - 2 120 59

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK142052.1, X95503.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 429547 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0