This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00494693_m1
See other GOT1 GE Assays ›
Gene Symbol:
Got1
Gene Name:
glutamic-oxaloacetic transaminase 1, soluble
Gene Aliases:
AI789014, Got-1, cAspAT, cCAT
Chromosome Location:
Chr.19: 43499752 - 43524605 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00494693_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Got1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010324.2 NP_034454.2 2 - 3 449 87
GenBank mRNA AK146445.1 - 2 - 3 351 87
AK147934.1 - 2 - 3 354 87
AK192547.1 - 1 - 2 59 87
BC002057.1 - 2 - 3 337 3489628 87
CK334037.1 - 2 - 3 449 87
J02623.1 - 2 - 3 353 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001541.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Got1

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

oxaloacetate metabolic process
glycerol biosynthetic process
cellular amino acid metabolic process
aspartate metabolic process
aspartate biosynthetic process
aspartate catabolic process
Notch signaling pathway
cellular amino acid biosynthetic process
biosynthetic process
glutamate catabolic process to aspartate
glutamate catabolic process to 2-oxoglutarate
dicarboxylic acid metabolic process
fatty acid homeostasis
catalytic activity
L-aspartate:2-oxoglutarate aminotransferase activity
phosphatidylserine decarboxylase activity
transaminase activity
transferase activity
pyridoxal phosphate binding
carboxylic acid binding
L-cysteine:2-oxoglutarate aminotransferase activity
L-phenylalanine:2-oxoglutarate aminotransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900013594.mm.1 NM_010324.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013594.mm.1 NM_010324.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900013594.mm.1 TC1900001541.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900013596.mm.1 NM_010324.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013596.mm.1 NM_010324.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900013596.mm.1 TC1900001541.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900013598.mm.1 NM_010324.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013598.mm.1 NM_010324.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900013598.mm.1 TC1900001541.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900013600.mm.1 NM_010324.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900013600.mm.1 NM_010324.2 Refseq
Affymetrix Mouse Clariom S Array PSR1900013600.mm.1 TC1900001541.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0