This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00494816_m1
See other GTF2I GE Assays ›
Gene Symbol:
Gtf2i
Gene Name:
general transcription factor II I
Gene Aliases:
6030441I21Rik, BAP-135, Diws1t, GtfII-I, Spin, TFII-I, WBSCR6
Chromosome Location:
Chr.5: 134237832 - 134314760 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00494816_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf2i
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080746.2 NP_001074215.1 6 - 7 969 122
NM_001080747.2 NP_001074216.1 6 - 7 969 122
NM_001080748.2 NP_001074217.1 6 - 7 969 122
NM_001080749.2 NP_001074218.1 6 - 7 969 122
NM_010365.4 NP_034495.2 6 - 7 969 122
GenBank mRNA AF017085.1 - 7 - 8 673 122
AF043219.1 - 5 - 6 590 122
AF043220.1 - 5 - 6 590 122
AK147201.1 - 6 - 7 927 122
AY030290.1 - 6 - 7 662 122
AY030291.1 - 6 - 7 662 122
AY030292.1 - 6 - 7 662 122
AY030293.1 - 6 - 7 662 122
BC047387.1 - 6 - 7 617 122
BC053044.1 - 6 - 7 938 5704935 122

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003369.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf2i

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
embryo development
transition between slow and fast fiber
negative regulation of angiogenesis
negative regulation of cytosolic calcium ion concentration
DNA binding
protein binding
mitogen-activated protein kinase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500030329.mm.1 TC0500003369.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500030329.mm.1 NM_001080746.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030329.mm.1 NM_001080747.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030329.mm.1 NM_001080748.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030329.mm.1 NM_001080749.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030329.mm.1 NM_010365.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0