This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00494829_m1
See other GTF2I GE Assays ›
Gene Symbol:
Gtf2i
Gene Name:
general transcription factor II I
Gene Aliases:
6030441I21Rik, BAP-135, Diws1t, GtfII-I, Spin, TFII-I, WBSCR6
Chromosome Location:
Chr.5: 134237832 - 134314760 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00494829_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf2i
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080746.2 NP_001074215.1 20 - 21 2134 85
NM_001080747.2 NP_001074216.1 19 - 20 2071 85
NM_001080748.2 NP_001074217.1 18 - 19 2014 85
NM_001080749.2 NP_001074218.1 18 - 19 2014 85
NM_010365.4 NP_034495.2 19 - 20 2077 85
GenBank mRNA AF017085.1 - 20 - 21 1778 85
AK145698.1 - 10 - 11 962 85
AK147201.1 - 18 - 19 1972 85
AY030290.1 - 19 - 20 1764 85
AY030291.1 - 19 - 20 1770 85
AY030292.1 - 18 - 19 1707 85
AY030293.1 - 20 - 21 1827 85
BC053044.1 - 20 - 21 2103 5704935 85

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf2i

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
embryo development
transition between slow and fast fiber
negative regulation of angiogenesis
negative regulation of cytosolic calcium ion concentration
DNA binding
protein binding
mitogen-activated protein kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0