This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00494833_m1
See other GTF2I GE Assays ›
Gene Symbol:
Gtf2i
Gene Name:
general transcription factor II I
Gene Aliases:
6030441I21Rik, BAP-135, Diws1t, GtfII-I, Spin, TFII-I, WBSCR6
Chromosome Location:
Chr.5: 134237832 - 134314760 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00494833_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf2i
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080746.2 NP_001074215.1 24 - 25 2552 61
NM_001080747.2 NP_001074216.1 23 - 24 2489 61
NM_001080748.2 NP_001074217.1 22 - 23 2432 61
NM_001080749.2 NP_001074218.1 22 - 23 2432 61
NM_010365.4 NP_034495.2 23 - 24 2495 61
GenBank mRNA AF017085.1 - 24 - 25 2196 61
AF043219.1 - 22 - 23 2116 61
AF043220.1 - 21 - 22 2053 61
AK145698.1 - 14 - 15 1380 61
AK147201.1 - 22 - 23 2390 61
AY030290.1 - 23 - 24 2182 61
AY030291.1 - 23 - 24 2188 61
AY030292.1 - 22 - 23 2125 61
AY030293.1 - 24 - 25 2245 61
BC047387.1 - 22 - 23 2080 61
BC053044.1 - 24 - 25 2521 5704935 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003369.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf2i

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
embryo development
transition between slow and fast fiber
negative regulation of angiogenesis
negative regulation of cytosolic calcium ion concentration
DNA binding
protein binding
mitogen-activated protein kinase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500030301.mm.1 TC0500003369.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500030301.mm.1 NM_001080746.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030301.mm.1 NM_001080747.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030301.mm.1 NM_001080748.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030301.mm.1 NM_001080749.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030301.mm.1 NM_010365.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-nbv7w:80/100.66.76.242:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0