This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00495330_g1
See other LIG1 GE Assays ›
Gene Symbol:
Lig1
Gene Name:
ligase I, DNA, ATP-dependent
Gene Aliases:
AL033288, LigI
Chromosome Location:
Chr.7: 13277313 - 13311427 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00495330_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lig1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001083188.1 NP_001076657.1 20 - 21 2187 69
NM_001199310.1 NP_001186239.1 20 - 21 2195 69
NM_010715.2 NP_034845.2 20 - 21 2112 69
GenBank mRNA AK153993.1 - 20 - 21 2074 69
AK160208.1 - 5 - 6 481 69
AK161793.1 - 13 - 14 1270 69
AK168643.1 - 20 - 21 2191 69
BC028287.1 - 20 - 21 2037 4911261 69
BC138541.1 - 20 - 21 2195 8861561 69
BC138542.1 - 20 - 21 2195 8861562 69
U04674.1 - 19 - 20 1980 69
U19604.1 - 20 - 21 2225 69

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lig1

Panther Classification:

Molecular Function -

DNA ligase

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
DNA ligation
lagging strand elongation
DNA repair
base-excision repair
double-strand break repair via nonhomologous end joining
DNA recombination
cellular response to DNA damage stimulus
cell cycle
response to hydrogen peroxide
DNA ligation involved in DNA repair
cell division
DNA biosynthetic process
Okazaki fragment processing involved in mitotic DNA replication
removal of RNA primer involved in mitotic DNA replication
nucleotide binding
DNA binding
DNA ligase activity
DNA ligase (ATP) activity
ATP binding
ligase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-kt9qp:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0