This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00495353_m1
See other LIPE GE Assays ›
Gene Symbol:
Lipe
Gene Name:
lipase, hormone sensitive
Gene Aliases:
4933403G17Rik, HSL
Chromosome Location:
Chr.7: 25379527 - 25398704 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00495353_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lipe
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039507.2 NP_001034596.1 2 - 3 581 90
NM_010719.5 NP_034849.2 2 - 3 1017 90
GenBank mRNA AK019846.1 - 2 - 3 1418 90
AK029984.1 - 2 - 3 1020 90
AK169858.1 - 2 - 3 576 90
BC021642.1 - 2 - 3 577 4188567 90
BC053502.1 - 2 - 3 641 90
U08188.1 - 1 - 2 1102 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002632.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lipe

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

transcription initiation from RNA polymerase I promoter
termination of RNA polymerase I transcription
lipid metabolic process
metabolic process
steroid metabolic process
cholesterol metabolic process
lipid catabolic process
triglyceride catabolic process
long-chain fatty acid catabolic process
diacylglycerol catabolic process
triglyceride lipase activity
protein binding
lipase activity
hydrolase activity
hydrolase activity, acting on ester bonds
protein kinase binding
hormone-sensitive lipase activity
rRNA primary transcript binding
acylglycerol lipase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700022751.mm.1 TC0700002632.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700022751.mm.1 NM_001039507.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700022751.mm.1 NM_010719.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700022751.mm.1 NM_010719.5 Refseq
Affymetrix Mouse Clariom S Array PSR0700022750.mm.1 TC0700002632.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700022750.mm.1 NM_001039507.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700022750.mm.1 NM_010719.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700022750.mm.1 NM_010719.5 Refseq
Affymetrix Mouse Clariom S Array PSR0700022749.mm.1 TC0700002632.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700022749.mm.1 NM_001039507.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700022749.mm.1 NM_010719.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700022749.mm.1 NM_010719.5 Refseq
Affymetrix Mouse Clariom S Array PSR0700022757.mm.1 TC0700002632.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700022757.mm.1 NM_010719.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700022757.mm.1 NM_010719.5 Refseq
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 793941 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0