This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00495357_g1
See other LIPE GE Assays ›
Gene Symbol:
Lipe
Gene Name:
lipase, hormone sensitive
Gene Aliases:
4933403G17Rik, HSL
Chromosome Location:
Chr.7: 25379527 - 25398704 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00495357_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lipe
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039507.2 NP_001034596.1 6 - 7 1300 62
NM_010719.5 NP_034849.2 6 - 7 1736 62
GenBank mRNA AK019846.1 - 6 - 7 2137 62
AK029984.1 - 6 - 7 1739 62
AK169858.1 - 6 - 7 1295 62
BC021642.1 - 6 - 7 1296 4188567 62
BC053502.1 - 6 - 7 1360 62
U08188.1 - 5 - 6 1821 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002632.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lipe

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

transcription initiation from RNA polymerase I promoter
termination of RNA polymerase I transcription
lipid metabolic process
metabolic process
steroid metabolic process
cholesterol metabolic process
lipid catabolic process
triglyceride catabolic process
long-chain fatty acid catabolic process
diacylglycerol catabolic process
triglyceride lipase activity
protein binding
lipase activity
hydrolase activity
hydrolase activity, acting on ester bonds
protein kinase binding
hormone-sensitive lipase activity
rRNA primary transcript binding
acylglycerol lipase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700022748.mm.1 TC0700002632.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700022748.mm.1 NM_001039507.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700022748.mm.1 NM_010719.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700022748.mm.1 NM_010719.5 Refseq
Affymetrix Mouse Clariom S Array PSR0700022747.mm.1 TC0700002632.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700022747.mm.1 NM_001039507.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700022747.mm.1 NM_010719.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700022747.mm.1 NM_010719.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0