This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00495362_m1
See other LIPG GE Assays ›
Gene Symbol:
Lipg
Gene Name:
lipase, endothelial
Gene Aliases:
3110013K01Rik, EL, lipase, mEDL
Chromosome Location:
Chr.18: 74939322 - 74961263 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00495362_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lipg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010720.3 NP_034850.3 2 - 3 575 70
GenBank mRNA AF118768.1 - 2 - 3 560 70
AK090224.1 - 2 - 3 573 70
AK152161.1 - 2 - 3 577 70
AK152430.1 - 2 - 3 573 70
AK152872.1 - 2 - 3 575 70
AK153541.1 - 2 - 3 577 70
AK172101.1 - 2 - 3 575 70
BC020991.1 - 2 - 3 429 3981856 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001595.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lipg

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
cell proliferation
positive regulation of high-density lipoprotein particle clearance
lipid catabolic process
positive regulation of cholesterol transport
high-density lipoprotein particle remodeling
cholesterol homeostasis
reverse cholesterol transport
regulation of lipoprotein metabolic process
phospholipid homeostasis
lipoprotein lipase activity
phospholipase activity
triglyceride lipase activity
heparin binding
phosphatidylcholine 1-acylhydrolase activity
hydrolase activity
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800011518.mm.1 NM_010720.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800011518.mm.1 NM_010720.3 Refseq
Affymetrix Mouse Clariom S Array PSR1800011518.mm.1 TC1800001595.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0