This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00495694_m1
See other TOM1 GE Assays ›
Gene Symbol:
Tom1
Gene Name:
target of myb1 homolog (chicken)
Gene Aliases:
-
Chromosome Location:
Chr.8: 75033686 - 75070121 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00495694_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tom1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136259.1 NP_001129731.1 8 - 9 990 75
NM_011622.3 NP_035752.1 8 - 9 990 75
GenBank mRNA AJ006972.1 - 8 - 9 967 75
AK028398.1 - 8 - 9 931 75
AK051245.1 - 8 - 9 972 75
AK150143.1 - 8 - 9 940 75
AK160403.1 - 8 - 9 936 75
BC021633.1 - 8 - 9 953 4208226 75
BC093520.1 - 8 - 9 918 2866013 75
BC157905.1 - 8 - 9 951 9088106 75
BC158042.1 - 8 - 9 951 9007343 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000844.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tom1

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
protein transport
protein binding
clathrin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800006249.mm.1 NM_001136259.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006249.mm.1 NM_001136259.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006249.mm.1 NM_011622.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800006249.mm.1 NM_011622.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800006249.mm.1 TC0800000844.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0