This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00495908_g1
See other UCP2 GE Assays ›
Gene Symbol:
Ucp2
Gene Name:
uncoupling protein 2 (mitochondrial, proton carrier)
Gene Aliases:
Slc25a8
Chromosome Location:
Chr.7: 100493358 - 100499630 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00495908_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ucp2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011671.5 NP_035801.3 6 - 7 999 67
GenBank mRNA AF111998.1 - 4 - 5 702 67
AF111999.1 - 4 - 5 696 67
AK035298.1 - 994 67
AK054060.1 - 1000 67
AK134116.1 - 6 - 7 1000 67
AK142687.1 - 6 - 7 1000 67
AK165245.1 - 6 - 7 1000 67
BC012697.1 - 6 - 7 998 4205625 67
BC012967.1 - 5 - 6 900 2811421 67
U69135.1 - 6 - 7 927 67
U94593.1 - 6 - 7 920 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001409.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ucp2

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s)

response to superoxide
response to hypoxia
transport
mitochondrial transport
positive regulation of cell death
negative regulation of apoptotic process
regulation of mitochondrial membrane potential
negative regulation of insulin secretion involved in cellular response to glucose stimulus

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700012042.mm.1 TC0700001409.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700012042.mm.1 NM_011671.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0