This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00495934_g1
See other UNC119 GE Assays ›
Gene Symbol:
Unc119
Gene Name:
unc-119 homolog (C. elegans)
Gene Aliases:
D11Bhm52, HRG4, MRG4, Rg4, Rtg4, Unc119h
Chromosome Location:
Chr.11: 78343443 - 78349164 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00495934_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Unc119
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011676.2 NP_035806.1 3 - 4 486 61
GenBank mRNA AF030169.1 - 3 - 4 510 61
AK131955.1 - 3 - 4 491 61
BC001990.1 - 3 - 4 486 3484396 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001158.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Unc119

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

transport
endocytosis
visual perception
protein transport
lipoprotein transport
response to stimulus
positive regulation of protein tyrosine kinase activity
negative regulation of clathrin-mediated endocytosis
negative regulation of caveolin-mediated endocytosis
protein binding
lipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100010799.mm.1 NM_011676.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100010799.mm.1 TC1100001158.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100010798.mm.1 NM_011676.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100010798.mm.1 TC1100001158.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0