This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00495935_g1
See other UNC119 GE Assays ›
Gene Symbol:
Unc119
Gene Name:
unc-119 homolog (C. elegans)
Gene Aliases:
D11Bhm52, HRG4, MRG4, Rg4, Rtg4, Unc119h
Chromosome Location:
Chr.11: 78343443 - 78349164 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00495935_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Unc119
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011676.2 NP_035806.1 4 - 5 652 67
GenBank mRNA AF030169.1 - 4 - 5 676 67
AK131955.1 - 4 - 5 657 67
BC001990.1 - 4 - 5 652 3484396 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100001158.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Unc119

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

transport
endocytosis
visual perception
protein transport
lipoprotein transport
response to stimulus
positive regulation of protein tyrosine kinase activity
negative regulation of clathrin-mediated endocytosis
negative regulation of caveolin-mediated endocytosis
protein binding
lipid binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27942760 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27942765 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27942763 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27942764 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27942762 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27942761 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957378 BC001990.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957378 NM_011676.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1957378 AK131955.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 53957 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 231073 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 771538 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100369 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957378 BC001990.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957378 NM_011676.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1957378 AK131955.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201638 BC001990.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201638 NM_011676.2 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201638 AF030169.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P201638 AK131955.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27942781 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27942779 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27942775 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27942766 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27942784 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27942773 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27942770 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27942758 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27942762 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27942760 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27942778 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27942764 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100369 - -
Whole Mouse Genome G4122A A_51_P201638 BC001990.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P201638 NM_011676.2 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P201638 AF030169.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P201638 AK131955.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105474 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100369 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09661601 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09089927 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27942763 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27942760 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27942762 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27942764 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27942765 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27942761 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942759 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942825 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942826 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942816 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942810 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942817 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942793 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942805 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942770 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942798 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942832 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942796 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942820 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942787 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942781 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942758 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942785 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942806 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942827 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942775 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942797 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942766 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942801 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942791 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942779 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942803 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942789 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942795 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942764 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942773 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942784 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942778 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942762 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942760 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942818 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27942814 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942819 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942805 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942810 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942801 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942759 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942817 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942779 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942773 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942785 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09676082 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942788 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942770 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09307341 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942778 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09307347 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942806 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942818 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942760 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942766 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942816 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942793 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942784 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942781 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942798 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942789 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942822 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942803 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942814 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942797 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942757 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942758 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942791 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P09307349 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27942775 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10379177 - -
Affymetrix Mouse Gene 1.0 ST Array 10379182 - -
Affymetrix Mouse Gene 1.0 ST Array 10379179 - -
Affymetrix Mouse Gene 1.0 ST Array 10379180 - -
Affymetrix Mouse Gene 1.0 ST Array 10379183 - -
Affymetrix Mouse Gene 1.0 ST Array 10379178 - -
Affymetrix Mouse Gene 1.0 ST Array 10379181 - -
Affymetrix HT MG-430 PM Array Plate 1418123_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10379176 - -
Mouse Genome 430 2.0 Array 1418123_at - -
Mouse Genome 430 2.0 Array 1418123_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 22248_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 22248_PM_at - -
Affymetrix Mouse Clariom S Array PSR1100010804.mm.1 NM_011676.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100010804.mm.1 TC1100001158.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100010802.mm.1 NM_011676.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100010802.mm.1 TC1100001158.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100010801.mm.1 NM_011676.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100010801.mm.1 TC1100001158.mm.1 MTA TC
Mouse Expression Set 430 1418123_PM_at - -
Mouse Expression Set 430 1418123_at - -
Murine Genome U74v2 Set 95555_PM_at - -
Murine Genome U74v2 Set 95555_at - -
Mouse Genome 430A 2.0 Array 1418123_at - -
Mouse Genome 430A 2.0 Array 1418123_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1418123_at - -
Affymetrix HT Mouse Genome 430A Array 1418123_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 335834 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 360372 - -
Illumina mouse-6 v1.0 expression beadchip 360372 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000360372 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2464999 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2464999 - -
mouse-6 v1.1 (Illumina) scl41204.5.67_14-S - -
Illumina Sentrix mouse-6.v1 bead array 360372 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl41204.5.67_14-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 360372 - -
MouseRef-8 v2 Expression BeadChip ILMN_2847834 - -
MouseRef-8 v2 Expression BeadChip ILMN_2464999 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2464999 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2847834 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0