This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00496453_m1
See other ABCD2 GE Assays ›
Gene Symbol:
Abcd2
Gene Name:
ATP-binding cassette, sub-family D (ALD), member 2
Gene Aliases:
ABC39, ALDL1, ALDR, ALDRP
Chromosome Location:
Chr.15: 91145871 - 91192504 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00496453_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Abcd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011994.2 NP_036124.2 7 - 8 1992 78
GenBank mRNA AK051445.1 - 7 - 8 1966 78
AK082588.1 - 7 - 8 1964 78
AK087653.1 - 7 - 8 1611 78
AK134763.1 - 2 - 3 303 78
AK161027.1 - 7 - 8 1653 78
BC019187.1 - 7 - 8 1936 5027149 78
Z48670.1 - 7 - 8 1996 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002075.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Abcd2

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

very long-chain fatty acid metabolic process
fatty acid beta-oxidation
transport
positive regulation of fatty acid beta-oxidation
very long-chain fatty acid catabolic process
transmembrane transport
nucleotide binding
ATP binding
ATPase activity
ATPase activity, coupled to transmembrane movement of substances
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500016758.mm.1 NM_011994.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016758.mm.1 NM_011994.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500016758.mm.1 TC1500002075.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500016759.mm.1 NM_011994.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500016759.mm.1 NM_011994.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500016759.mm.1 TC1500002075.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline