This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00496576_m1
See other GPR162 GE Assays ›
Gene Symbol:
Gpr162
Gene Name:
G protein-coupled receptor 162
Gene Aliases:
A-2, BC031437, Grca
Chromosome Location:
Chr.6: 124858444 - 124864219 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00496576_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr162
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013533.3 NP_038561.1 1 - 2 247 106
GenBank mRNA AK134941.1 - 1 - 2 61 106
AK144543.1 - 1 - 2 34 106
BC031437.1 - 1 - 2 247 4527410 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600003123.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK134941.1,AK144543.1,BC031437.1,NM_013533.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr162

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
biological_process
molecular_function
signal transducer activity
G-protein coupled receptor activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600024346.mm.1 TC0600003123.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600024346.mm.1 NM_013533.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600024344.mm.1 TC0600003123.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600024344.mm.1 NM_013533.4 Refseq
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 559958 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0