This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00496578_g1
See other GPR162 GE Assays ›
Gene Symbol:
Gpr162
Gene Name:
G protein-coupled receptor 162
Gene Aliases:
A-2, BC031437, Grca
Chromosome Location:
Chr.6: 124858444 - 124864219 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00496578_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr162
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013533.3 NP_038561.1 3 - 4 1741 67
GenBank mRNA AK134941.1 - 3 - 4 1555 67
AK144354.1 - 2 - 3 1510 67
AK171145.1 - 4 - 5 1925 67
BC031437.1 - 3 - 4 1741 4527410 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600003123.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr162

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
biological_process
molecular_function
signal transducer activity
G-protein coupled receptor activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24817421 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24817444 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24817442 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24817423 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24817441 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24817420 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344376 BC031437.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344376 AK171145.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344376 BC016413.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344376 AK144543.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344376 AK158847.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344376 BC031437.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344376 AK171145.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344376 BC016413.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344376 AK144543.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P344376 AK158847.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 952946 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 788429 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 788627 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 942346 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 149635 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142006 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344376 BC031437.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344376 AK171145.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344376 BC016413.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344376 AK144543.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344376 AK158847.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344376 BC031437.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344376 AK171145.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344376 BC016413.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344376 AK144543.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P344376 AK158847.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P344376 BC031437.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P344376 AK171145.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P344376 BC016413.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P344376 AK144543.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P344376 AK158847.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P344376 BC031437.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P344376 AK171145.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P344376 BC016413.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P344376 AK144543.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P344376 AK158847.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817404 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817447 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817406 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817428 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817442 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817413 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817412 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817437 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817408 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817444 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817418 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817399 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817448 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817416 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817439 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817409 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817419 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817441 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817427 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817410 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817403 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817431 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817430 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817434 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817405 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817436 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817401 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817397 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817424 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817423 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817415 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817407 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817433 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24817426 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142006 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01437864 - -
Whole Mouse Genome G4122A A_51_P344376 BC031437.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P344376 AK171145.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P344376 BC016413.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P344376 AK144543.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P344376 AK158847.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P344376 BC031437.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P344376 AK171145.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P344376 BC016413.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P344376 AK144543.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P344376 AK158847.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115913 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142006 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05250538 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05250532 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05686224 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05686222 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24817421 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24817420 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24817444 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24817423 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24817441 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24817442 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817448 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817445 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817345 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817447 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817388 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817391 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817413 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817436 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817441 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817423 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817338 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817419 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817420 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817442 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817368 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817401 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817370 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817399 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817431 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817397 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817361 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817437 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817405 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817342 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817393 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817357 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817366 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817407 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817349 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817385 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817392 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817390 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817346 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817383 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817410 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817404 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817371 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817381 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817434 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817433 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817376 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817424 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817412 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817395 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817416 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817351 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817344 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817369 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817409 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817444 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817427 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817439 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817418 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817379 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817403 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817430 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24817406 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05250569 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05250582 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05594483 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05250540 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05250549 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05250535 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24817399 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05250548 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24817416 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24817447 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24817404 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05250533 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05250532 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05250570 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24817442 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24817445 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05250567 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24817430 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05250552 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05250529 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05250572 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24817406 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24817424 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05250530 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24817405 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05686222 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24817436 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05250556 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24817403 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05250547 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05250531 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05250566 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10547893 - -
Affymetrix Mouse Gene 1.0 ST Array 10547889 - -
Affymetrix Mouse Gene 1.0 ST Array 10547892 - -
Affymetrix Mouse Gene 1.0 ST Array 10547890 - -
Affymetrix Mouse Gene 1.0 ST Array 10547891 - -
Affymetrix HT MG-430 PM Array Plate 1418611_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10547888 - -
Mouse Genome 430 2.0 Array 1418611_PM_at - -
Mouse Genome 430 2.0 Array 1418611_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 14788_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 14788_at - -
Affymetrix Mouse Clariom S Array PSR0600024342.mm.1 TC0600003123.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600024342.mm.1 NM_013533.4 Refseq
Mouse Expression Set 430 1418611_at - -
Mouse Expression Set 430 1418611_PM_at - -
Murine Genome U74v2 Set 103931_at - -
Murine Genome U74v2 Set 161771_r_at - -
Murine Genome U74v2 Set 161771_PM_r_at - -
Murine Genome U74v2 Set 103931_PM_at - -
Mouse Genome 430A 2.0 Array 1418611_at - -
Mouse Genome 430A 2.0 Array 1418611_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1418611_at - -
Affymetrix HT Mouse Genome 430A Array 1418611_PM_at - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 730072 - -
MouseWG-6 v2 Expression BeadChip 7050402 - -
Illumina mouse-6 v1.0 expression beadchip 7050402 - -
Illumina mouse-6 v1.0 expression beadchip 730072 - -
Illumina mouseRef-8 v1.0 expression beadchip 0007050402 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000730072 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2700785 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2704110 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2704110 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2700785 - -
mouse-6 v1.1 (Illumina) scl0001152.1_862-S - -
mouse-6 v1.1 (Illumina) scl28414.13.1_30-S - -
Illumina Sentrix mouse-6.v1 bead array 730072 - -
Illumina Sentrix mouse-6.v1 bead array 7050402 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl28414.13.1_30-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0001152.1_862-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 7050402 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 730072 - -
MouseRef-8 v2 Expression BeadChip ILMN_2984942 - -
MouseRef-8 v2 Expression BeadChip ILMN_2704110 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2704110 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2984942 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0001152.1_862-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f5rhb:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0