This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00496965_g1
See other GCAT GE Assays ›
Gene Symbol:
Gcat
Gene Name:
glycine C-acetyltransferase (2-amino-3-ketobutyrate-coenzyme A ligase)
Gene Aliases:
AI526977, Kbl
Chromosome Location:
Chr.15: 79030874 - 79042531 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00496965_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gcat
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001161712.1 NP_001155184.1 4 - 5 630 83
NM_013847.4 NP_038875.3 4 - 5 630 83
GenBank mRNA AF093403.1 - 4 - 5 588 83
AK013138.1 - 4 - 5 585 83
AK075782.1 - 4 - 5 570 83
AK076469.1 - 4 - 5 603 83
AK087433.1 - 4 - 5 589 83
BC024107.1 - 4 - 5 630 5125361 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000663.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gcat

Panther Classification:

Molecular Function -

transaminase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500004927.mm.1 NM_001161712.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004927.mm.1 NM_001161712.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004927.mm.1 NM_013847.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500004927.mm.1 NM_013847.4 Refseq
Affymetrix Mouse Clariom S Array PSR1500004927.mm.1 TC1500000663.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0