This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00497405_g1
See other DLG3 GE Assays ›
Gene Symbol:
Dlg3
Gene Name:
discs, large homolog 3 (Drosophila)
Gene Aliases:
Dlgh3, SAP102, mKIAA1232
Chromosome Location:
Chr.X: 100767586 - 100818410 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00497405_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dlg3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177778.2 NP_001171249.1 12 - 13 2098 71
NM_001177779.1 NP_001171250.1 5 - 6 999 71
NM_001177780.1 NP_001171251.1 3 - 4 391 71
NM_001290402.1 NP_001277331.1 11 - 12 2044 71
NM_016747.4 NP_058027.1 12 - 13 2098 71
GenBank mRNA AK122474.1 - 12 - 13 2097 71
AK145815.1 - 5 - 6 754 71
AK148464.1 - 3 - 4 370 71
AK159655.1 - 3 - 4 379 71
AK167365.1 - 3 - 4 378 71
BC060713.1 - 10 - 11 2076 71
BC094368.1 - 11 - 12 2039 6842105 71
CN698511.1 - 3 - 4 391 71
D87117.1 - 12 - 13 2072 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00000963.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dlg3

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

establishment of planar polarity
chemical synaptic transmission
nervous system development
negative regulation of phosphatase activity
receptor clustering
establishment or maintenance of epithelial cell apical/basal polarity
nucleotide phosphorylation
receptor localization to synapse
guanylate kinase activity
protein binding
protein C-terminus binding
kinase binding
phosphatase binding
protein phosphatase binding
protein domain specific binding
PDZ domain binding
ubiquitin protein ligase binding
ionotropic glutamate receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00006299.mm.1 NM_001177778.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006299.mm.1 NM_001177779.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006299.mm.1 NM_001177779.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006299.mm.1 NM_001177780.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006299.mm.1 NM_001177780.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006299.mm.1 NM_016747.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006299.mm.1 TC0X00000963.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00006301.mm.1 NM_001177779.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006301.mm.1 NM_001177779.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006301.mm.1 NM_001177780.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006301.mm.1 NM_001177780.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006301.mm.1 TC0X00000963.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00006298.mm.1 NM_001177778.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006298.mm.1 NM_001177779.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006298.mm.1 NM_001177779.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006298.mm.1 NM_001177780.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006298.mm.1 NM_001177780.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006298.mm.1 NM_016747.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00006298.mm.1 TC0X00000963.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0