This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00498824_m1
See other LIPA GE Assays ›
Gene Symbol:
Lipa
Gene Name:
lysosomal acid lipase A
Gene Aliases:
AA960673, Lal, Lip-1, Lip1
Chromosome Location:
Chr.19: 34492316 - 34527474 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00498824_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lipa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001111100.1 NP_001104570.1 6 - 7 775 84
NM_021460.3 NP_067435.3 6 - 7 779 84
GenBank mRNA AK088659.1 - 6 - 7 731 84
AK149791.1 - 6 - 7 739 84
AK154804.1 - 6 - 7 721 84
AK164007.1 - 6 - 7 775 84
AK169567.1 - 6 - 7 745 84
BC058564.1 - 6 - 7 727 6441278 84
Z31689.1 - 5 - 6 684 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900001419.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lipa

Panther Classification:

Molecular Function -

hydrolase lipase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
cytokine production
lipid metabolic process
fatty acid metabolic process
inflammatory response
cell proliferation
lipid catabolic process
sterol metabolic process
lung development
tissue remodeling
homeostasis of number of cells within a tissue
sterol esterase activity
lipase activity
hydrolase activity
hydrolase activity, acting on ester bonds

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0