This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00499082_m1
See other ITM2C GE Assays ›
Gene Symbol:
Itm2c
Gene Name:
integral membrane protein 2C
Gene Aliases:
3110038L02Rik, BRI3, Bricd2c, E25, E25C, ITM3
Chromosome Location:
Chr.1: 85894510 - 85908698 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00499082_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Itm2c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022417.3 NP_071862.2 3 - 4 611 66
GenBank mRNA AB030199.1 - 3 - 4 554 66
AF272044.1 - 3 - 4 518 66
AF282981.1 - 3 - 4 539 66
AK036377.1 - 3 - 4 557 66
AK156930.1 - 3 - 4 514 66
BC012952.1 - 3 - 4 489 2648812 66

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Itm2c

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of neuron projection development
neuron differentiation
positive regulation of extrinsic apoptotic signaling pathway
beta-amyloid binding
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0