This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00499533_m1
See other DGAT2 GE Assays ›
Gene Symbol:
Dgat2
Gene Name:
diacylglycerol O-acyltransferase 2
Gene Aliases:
0610010B06Rik, ARAT, DGAT-2
Chromosome Location:
Chr.7: 99153663 - 99182713 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00499533_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dgat2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026384.3 NP_080660.1 4 - 5 637 61
GenBank mRNA AF384160.1 - 4 - 5 495 61
AK002443.1 - 4 - 5 642 61
AK016660.1 - 4 - 5 585 61
BC043447.1 - 4 - 5 637 5125951 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700003793.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dgat2

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycerol metabolic process
lipid metabolic process
triglyceride biosynthetic process
lipid storage
low-density lipoprotein particle clearance
long-chain fatty-acyl-CoA metabolic process
cellular triglyceride homeostasis
cholesterol homeostasis
diacylglycerol metabolic process
fatty acid homeostasis
fat pad development
cellular response to oleic acid
regulation of plasma lipoprotein particle levels
2-acylglycerol O-acyltransferase activity
diacylglycerol O-acyltransferase activity
transferase activity
transferase activity, transferring acyl groups
transferase activity, transferring acyl groups other than amino-acyl groups
protein homodimerization activity
retinol O-fatty-acyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0