This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00499549_m1
See other ATP5S GE Assays ›
Gene Symbol:
Atp5s
Gene Name:
ATP synthase, H+ transporting, mitochondrial F0 complex, subunit S
Gene Aliases:
1110015E18Rik, ATPW
Chromosome Location:
Chr.12: 69724930 - 69744665 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00499549_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5s
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026536.1 NP_080812.1 1 - 2 52 67
GenBank mRNA AK003725.1 - 1 - 2 52 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200000576.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK003725.1,NM_026536.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5s

Panther Classification:

Molecular Function -

ATP synthase primary active transporter transporter

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
transport
ion transport
proton transport
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200003851.mm.1 NM_026536.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003851.mm.1 NM_026536.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003851.mm.1 TC1200000576.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1200003848.mm.1 NM_026536.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003848.mm.1 NM_026536.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200003848.mm.1 TC1200000576.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 416391 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0