This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00499983_m1
See other CDHR1 GE Assays ›
Gene Symbol:
Cdhr1
Gene Name:
cadherin-related family member 1
Gene Aliases:
Pcdh21, Prcad, mKIAA1775
Chromosome Location:
Chr.14: 37077849 - 37098311 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00499983_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdhr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_130878.2 NP_570948.1 5 - 6 609 77
GenBank mRNA AB093300.1 - 5 - 6 1025 77
AF426393.1 - 5 - 6 496 77
AK143151.1 - 5 - 6 548 77
BC042454.1 - 5 - 6 519 5369576 77
BC058270.1 - 5 - 6 609 6490597 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400001745.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdhr1

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
cellular process
photoreceptor cell maintenance
calcium ion binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400013589.mm.1 NM_130878.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400013589.mm.1 NM_130878.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400013589.mm.1 TC1400001745.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline